Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Buffers
Cloning Buffers
VWR® Ultra Pure Grade, Sm buffer
VWR® Ultra Pure Grade, Sm buffer
VWR® Ultra Pure Grade, Sm buffer
Catalog # J614-500ML
Supplier:
VWR Chemicals
CAS Number:
Certificates
Expand image
VWR® Ultra Pure Grade, Sm buffer
Catalog # J614-500ML
Supplier:
VWR Chemicals
CAS Number:
Certificates
Specifications
Pk:
500 mL
Description:
SM BUFFER
pH value:
7,40 - 7,60
Specifications
About this item
Product Details
Documentation
Clear haze-free solution
DNase-free
Product Details
Documentation