VWR® ONEBAG®, Dehydrated buffered peptone water in bags with reconstitution water
Ready to use buffered peptone water is prepared a few minutes before use, to guarantee the best performance of the medium.
- Long shelf life: 24 months at room temperature
- Bag, water and culture medium sterilised by gamma irradiation (no over-heating; better performance of the medium)
- No need for dispensing equipment (eg. diluter, deionised water producer and more)
- Culture media is prepared in a few minutes before use, to guaranteee the best performance of the medium
Available to make up 90 or 225 ml of medium.
: In compliance with ISO 6887, ISO 6579 and ISO 11290-2.