Order Entry
Germany
ContactUsLinkComponent
Analytical standard, Diquat dibromid Monohydrate, reference material, Fluka™
Analytical standard, Diquat dibromid Monohydrate, reference material, Fluka™
Catalog # HONCR1775-100MG
CAS Number:  
Analytical standard, Diquat dibromid Monohydrate, reference material, Fluka™
Catalog # HONCR1775-100MG
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 mg
  • Standards:
    Reference material
  • CAS No.:
    6385-62-2
  • Description:
    Diquat dibromid Monohydrate
  • Formula:
    C12H12Br2N2 · H2O
  • Description:
    Diquat dibromid Monohydrate
  • Form:
    Neat

Specifications

About this item