Thiocyanate standard for ion chromatography, (CRM), Fluka™
Catalog # HONCE3050-100ML
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Concentration:1000 mg/L
- Matrix:H₂O
- Pack type:Glass bottle
- Pk:100 mL
- Description:Thiocyanate standard for IC
Specifications
About this item
Certified Reference Material (CRM)
- Traceable to SRM from NIST
Certifications: ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB).