Concentrated Eluent for Ion Chromatography, Sodium Carbonate Solution
Catalog # HONCE2206-500ML
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Matrix:H₂O
- Pack type:Plastic bottle
- Pk:500 mL
- Description:Concentrated Eluent for Ion Chromatography, Sodium Carbonate Solution
Specifications
About this item
Certified Reference Material (CRM).
0.5 M Sodium Carbonate (Na₂CO₃) in water