Order Entry
Germany
ContactUsLinkComponent
Ammonium nitrate 50 g/L in water matrix modifier solution, for atomic absorption spectroscopy, Fluka™
Catalog # HONCE4604-50ML
CAS Number:  
Ammonium nitrate 50 g/L in water matrix modifier solution, for atomic absorption spectroscopy, Fluka™
Catalog # HONCE4604-50ML
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    50 mL
  • Seq:
    2370

Specifications

Specification Test Results

  • Matrix:
    H₂O
  • Concentration:
    NH₄NO₃ 50g/L
  • Quality:
    ISO/IEC17025:2017(ANAB),ISO17034:2016(ANAB)
  • Form:
    Solution
  • Density:
    1.01g/cm3

Specification Test Results

About this item

 :
 :80.04 g/mol
Density:1.01 g/cm3
Storage Temperature:Ambient
CAS Number:6484-52-2
Formula:NH₄NO₃
MDL Number:MFCD00011425