Order Entry
Germany
ContactUsLinkComponent
Thermoshaker, Matrix Orbital
Thermoshaker, Matrix Orbital
Catalog # 444-1277
Supplier:  IKA
Thermoshaker, Matrix Orbital
Catalog # 444-1277
Supplier:  IKA

Specifications

  • Orbit:
    3 mm
  • Description:
    Thermoshaker, IKA Matrix Orbital, EU-plug
  • Seq:
    01
  • Model:
    IKA Matrix Orbital
  • Mixing speed range:
    0 - 3000 min⁻¹
  • Weight:
    7,2 kg
  • W×D×H:
    200×120×320 mm
  • Pk:
    1
  • Type:
    Orbital

Specifications

About this item

The IKA Matrix Orbital is part of a powerful product family of thermoshakers which have numerous user-friendly features.

  • Large, clear display with a convenient menu structure allowing intuitive operation
  • Speeds of up to 3000 rpm and shaking diameter of 3 mm ensure the liquid mixes quickly and effectively
  • Two different speed modes for optimal mixing
  • RS232 and USB interfaces

IKA Matrix Orbital mixes and is suitable foruse with variable inserts. Whether it is blood samples, pharmaceutical agents, DNA/RNA samples or ELISA assays – samples with even the smallest volumes mix reliably and completely in all laboratory applications. No cross-contamination and an optimal mixing result.

Certifications: Meets ASTM standards. EN-342. IP Protection Class according to DIN EN 60529: IP 21
Delivery: Supplied with inserts for 0,2 ml PCR vials and 0,5 and 1,5/2,0 ml reaction vials.