Albumin (from Egg white)
Catalog # TCIAA0198-25G
Supplier: TCI
CAS Number:
Specifications
- Pk:25 g
- Source:Egg white
- Storage conditions:Refrigerator
- Form:White to Brown powder to crystalline
- MDL No.:MFCD00130424
- Protein synonyms:Ovalbumin
- Protein/peptide name:Albumin