Order Entry
Germany
ContactUsLinkComponent
Avantor® Hichrom HI-5 MS, GC Columns
Avantor® Hichrom HI-5 MS, GC Columns
Supplier:  Avantor
Certificates
undefined
Avantor® Hichrom HI-5 MS, GC Columns
Supplier:  Avantor
Certificates

Specifications

  • Catalog No:
  • HI17-32-150-50
  • HI17-32-150-30
  • HI17-25-050-15
  • HI17-32-025-30G5
  • HI17-25-050-105
  • HI17-32-100-30G5
  • HI17-53-025-15
  • HI17-25-050-30G10
  • HI17-25-050-30G5
  • HI17-25-025-60
  • HI17-18-018-60
  • HI17-25-025-105
  • HI17-25-025-30G10
  • HI17-15-015-5
  • HI17-25-025-30G5
  • HI17-25-025-10
  • HI17-32-150-105
  • HI17-25-010-30G5
  • HI17-15-015-20
  • HI17-32-150-10
  • HI17-25-025-15G5
  • HI17-25-050-15G10
  • HI17-25-025-30
  • HI17-25-025-60G10
  • HI17-25-025-15
  • HI17-18-020-10
  • HI17-25-025-60G5
  • HI17-18-020-60
  • HI17-32-100-105
  • HI17-25-010-30
  • HI17-32-100-50
  • HI17-25-010-15
  • HI17-32-100-15
  • HI17-20-040-60
  • HI17-10-020-10
  • HI17-53-150-15
  • HI17-32-050-15
  • HI17-10-010-15
  • HI17-32-050-105
  • HI17-10-040-15
  • HI17-20-100-20
  • HI17-10-020-5
  • HI17-53-150-50
  • HI17-53-150-30
  • HI17-15-015-10
  • HI17-10-040-5
  • HI17-32-025-15
  • HI17-32-025-105
  • HI17-32-050-10
  • HI17-20-040-10
  • HI17-20-033-60
  • HI17-32-025-60
  • HI17-32-010-60
  • HI17-53-050-15
  • HI17-53-100-10
  • HI17-20-033-25
  • HI17-53-050-50
  • HI17-20-033-12
  • HI17-53-050-30
  • HI17-05-005-2.5
  • HI17-53-100-30
  • HI17-53-100-15
  • HI17-05-005-2
  • HI17-25-100-30
  • HI17-32-010-105
  • HI17-25-100-60
  • HI17-25-100-10
  • HI17-18-040-20
  • HI17-53-100-30G5
  • HI17-18-040-10
  • HI17-53-025-60
  • HI17-25-050-50
  • HI17-53-025-50
  • HI17-18-036-60
  • HI17-20-010-20
  • HI17-25-100-105
  • HI17-18-040-50
  • Phase name:
  • HI-5 MS
  • HI-5 MS
  • HI-5 MS
  • HI-5 MS
  • HI-5 MS
  • HI-5 MS
  • HI-5 MS
  • HI-5 MS
  • HI-5 MS
  • HI-5 MS
  • HI-5 MS
  • HI-5 MS
  • HI-5 MS
  • HI-5 MS
  • HI-5 MS
  • HI-5 MS
  • HI-5 MS
  • HI-5 MS
  • HI-5 MS
  • HI-5 MS
  • HI-5 MS
  • HI-5 MS
  • HI-5 MS
  • HI-5 MS
  • HI-5 MS
  • HI-5 MS
  • HI-5 MS
  • HI-5 MS
  • HI-5 MS
  • HI-5 MS
  • HI-5 MS
  • HI-5 MS
  • HI-5 MS
  • HI-5 MS
  • HI-5 MS
  • HI-5 MS
  • HI-5 MS
  • HI-5 MS
  • HI-5 MS
  • HI-5 MS
  • HI-5 MS
  • HI-5 MS
  • HI-5 MS
  • HI-5 MS
  • HI-5 MS
  • HI-5 MS
  • HI-5 MS
  • HI-5 MS
  • HI-5 MS
  • HI-5 MS
  • HI-5 MS
  • HI-5 MS
  • HI-5 MS
  • HI-5 MS
  • HI-5 MS
  • HI-5 MS
  • HI-5 MS
  • HI-5 MS
  • HI-5 MS
  • HI-5 MS
  • HI-5 MS
  • HI-5 MS
  • HI-5 MS
  • HI-5 MS
  • HI-5 MS
  • HI-5 MS
  • HI-5 MS
  • HI-5 MS
  • HI-5 MS
  • HI-5 MS
  • HI-5 MS
  • HI-5 MS
  • HI-5 MS
  • HI-5 MS
  • HI-5 MS
  • HI-5 MS
  • HI-5 MS
  • Phase composition:
  • 5% Phenyl, 95% Methyl Polysiloxane - low bleeding
  • 5% Phenyl, 95% Methyl Polysiloxane - low bleeding
  • 5% Phenyl, 95% Methyl Polysiloxane - low bleeding
  • 5% Phenyl, 95% Methyl Polysiloxane - low bleeding
  • 5% Phenyl, 95% Methyl Polysiloxane - low bleeding
  • 5% Phenyl, 95% Methyl Polysiloxane - low bleeding
  • 5% Phenyl, 95% Methyl Polysiloxane - low bleeding
  • 5% Phenyl, 95% Methyl Polysiloxane - low bleeding
  • 5% Phenyl, 95% Methyl Polysiloxane - low bleeding
  • 5% Phenyl, 95% Methyl Polysiloxane - low bleeding
  • 5% Phenyl, 95% Methyl Polysiloxane - low bleeding
  • 5% Phenyl, 95% Methyl Polysiloxane - low bleeding
  • 5% Phenyl, 95% Methyl Polysiloxane - low bleeding
  • 5% Phenyl, 95% Methyl Polysiloxane - low bleeding
  • 5% Phenyl, 95% Methyl Polysiloxane - low bleeding
  • 5% Phenyl, 95% Methyl Polysiloxane - low bleeding
  • 5% Phenyl, 95% Methyl Polysiloxane - low bleeding
  • 5% Phenyl, 95% Methyl Polysiloxane - low bleeding
  • 5% Phenyl, 95% Methyl Polysiloxane - low bleeding
  • 5% Phenyl, 95% Methyl Polysiloxane - low bleeding
  • 5% Phenyl, 95% Methyl Polysiloxane - low bleeding
  • 5% Phenyl, 95% Methyl Polysiloxane - low bleeding
  • 5% Phenyl, 95% Methyl Polysiloxane - low bleeding
  • 5% Phenyl, 95% Methyl Polysiloxane - low bleeding
  • 5% Phenyl, 95% Methyl Polysiloxane - low bleeding
  • 5% Phenyl, 95% Methyl Polysiloxane - low bleeding
  • 5% Phenyl, 95% Methyl Polysiloxane - low bleeding
  • 5% Phenyl, 95% Methyl Polysiloxane - low bleeding
  • 5% Phenyl, 95% Methyl Polysiloxane - low bleeding
  • 5% Phenyl, 95% Methyl Polysiloxane - low bleeding
  • 5% Phenyl, 95% Methyl Polysiloxane - low bleeding
  • 5% Phenyl, 95% Methyl Polysiloxane - low bleeding
  • 5% Phenyl, 95% Methyl Polysiloxane - low bleeding
  • 5% Phenyl, 95% Methyl Polysiloxane - low bleeding
  • 5% Phenyl, 95% Methyl Polysiloxane - low bleeding
  • 5% Phenyl, 95% Methyl Polysiloxane - low bleeding
  • 5% Phenyl, 95% Methyl Polysiloxane - low bleeding
  • 5% Phenyl, 95% Methyl Polysiloxane - low bleeding
  • 5% Phenyl, 95% Methyl Polysiloxane - low bleeding
  • 5% Phenyl, 95% Methyl Polysiloxane - low bleeding
  • 5% Phenyl, 95% Methyl Polysiloxane - low bleeding
  • 5% Phenyl, 95% Methyl Polysiloxane - low bleeding
  • 5% Phenyl, 95% Methyl Polysiloxane - low bleeding
  • 5% Phenyl, 95% Methyl Polysiloxane - low bleeding
  • 5% Phenyl, 95% Methyl Polysiloxane - low bleeding
  • 5% Phenyl, 95% Methyl Polysiloxane - low bleeding
  • 5% Phenyl, 95% Methyl Polysiloxane - low bleeding
  • 5% Phenyl, 95% Methyl Polysiloxane - low bleeding
  • 5% Phenyl, 95% Methyl Polysiloxane - low bleeding
  • 5% Phenyl, 95% Methyl Polysiloxane - low bleeding
  • 5% Phenyl, 95% Methyl Polysiloxane - low bleeding
  • 5% Phenyl, 95% Methyl Polysiloxane - low bleeding
  • 5% Phenyl, 95% Methyl Polysiloxane - low bleeding
  • 5% Phenyl, 95% Methyl Polysiloxane - low bleeding
  • 5% Phenyl, 95% Methyl Polysiloxane - low bleeding
  • 5% Phenyl, 95% Methyl Polysiloxane - low bleeding
  • 5% Phenyl, 95% Methyl Polysiloxane - low bleeding
  • 5% Phenyl, 95% Methyl Polysiloxane - low bleeding
  • 5% Phenyl, 95% Methyl Polysiloxane - low bleeding
  • 5% Phenyl, 95% Methyl Polysiloxane - low bleeding
  • 5% Phenyl, 95% Methyl Polysiloxane - low bleeding
  • 5% Phenyl, 95% Methyl Polysiloxane - low bleeding
  • 5% Phenyl, 95% Methyl Polysiloxane - low bleeding
  • 5% Phenyl, 95% Methyl Polysiloxane - low bleeding
  • 5% Phenyl, 95% Methyl Polysiloxane - low bleeding
  • 5% Phenyl, 95% Methyl Polysiloxane - low bleeding
  • 5% Phenyl, 95% Methyl Polysiloxane - low bleeding
  • 5% Phenyl, 95% Methyl Polysiloxane - low bleeding
  • 5% Phenyl, 95% Methyl Polysiloxane - low bleeding
  • 5% Phenyl, 95% Methyl Polysiloxane - low bleeding
  • 5% Phenyl, 95% Methyl Polysiloxane - low bleeding
  • 5% Phenyl, 95% Methyl Polysiloxane - low bleeding
  • 5% Phenyl, 95% Methyl Polysiloxane - low bleeding
  • 5% Phenyl, 95% Methyl Polysiloxane - low bleeding
  • 5% Phenyl, 95% Methyl Polysiloxane - low bleeding
  • 5% Phenyl, 95% Methyl Polysiloxane - low bleeding
  • 5% Phenyl, 95% Methyl Polysiloxane - low bleeding
  • Polarity:
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Crossbond:
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Temperature limit:
  • 350 °C
  • 350 °C
  • 350 °C
  • 350 °C
  • 350 °C
  • 350 °C
  • 350 °C
  • 350 °C
  • 350 °C
  • 350 °C
  • 350 °C
  • 350 °C
  • 350 °C
  • 350 °C
  • 350 °C
  • 350 °C
  • 350 °C
  • 350 °C
  • 350 °C
  • 350 °C
  • 350 °C
  • 350 °C
  • 350 °C
  • 350 °C
  • 350 °C
  • 350 °C
  • 350 °C
  • 350 °C
  • 350 °C
  • 350 °C
  • 350 °C
  • 350 °C
  • 350 °C
  • 350 °C
  • 350 °C
  • 350 °C
  • 350 °C
  • 350 °C
  • 350 °C
  • 350 °C
  • 350 °C
  • 350 °C
  • 350 °C
  • 350 °C
  • 350 °C
  • 350 °C
  • 350 °C
  • 350 °C
  • 350 °C
  • 350 °C
  • 350 °C
  • 350 °C
  • 350 °C
  • 350 °C
  • 350 °C
  • 350 °C
  • 350 °C
  • 350 °C
  • 350 °C
  • 350 °C
  • 350 °C
  • 350 °C
  • 350 °C
  • 350 °C
  • 350 °C
  • 350 °C
  • 350 °C
  • 350 °C
  • 350 °C
  • 350 °C
  • 350 °C
  • 350 °C
  • 350 °C
  • 350 °C
  • 350 °C
  • 350 °C
  • 350 °C
  • USP code:
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41

Specifications

Related Information

Frequently Bought Together