Certified thermometers with DKD certificate
Supplier: Amarell
Certificates
About this item
These precision thermometers, with enclosed scale, are suitable for government testing and/or official calibration.
- Total immersion
- Colourless capillary
- Laboratory milk glass
Filling: red spirit.
Ø: 8,0 - 9,0 mm
Length: 420 mm
Delivery: Supplied with a DKD certificate.