Order Entry
Germany
Orders LinkContactUsLinkComponent
Pyronin G, Sigma-Aldrich®

Pyronin G, Sigma-Aldrich®

Catalog #
Pyronin G, Sigma-Aldrich®
Catalog #
Supplier:  
CAS Number:  92-32-0
Pyronin G, Sigma-Aldrich®
Pyronin G, Sigma-Aldrich®
CAS Number:  92-32-0
Product Details & Documents
Synonyms:  Pyronin Y, Pyronine Y, 3,6-Bis(dimethylamino)xanthylium chloride
  • CAS Number:92-32-0
  • Formula:C₁₇H₁₉ClN₂O
  • MDL Number:MFCD00011725
  • Molecular Weight:302.8 g/mol
  • Merck Index:12,08190
  • Melting Pt:250…260 °C
  • Storage Temperature:Ambient
  • EINECS:202-147-8

Documents