Order Entry
Germany
Orders LinkContactUsLinkComponent
2-Hydroxy-5-nitropyridine

2-Hydroxy-5-nitropyridine

Glass bottle for solids, 100 g
Catalog # MOLE17835073-100G
Marketsource
Supplier:   Molekula
2-Hydroxy-5-nitropyridine
Catalog # MOLE17835073-100G
Supplier:  Molekula
Marketsource
CAS Number:  5418-51-9
2-Hydroxy-5-nitropyridine

Glass bottle for solids, 100 g

CAS Number:  5418-51-9
Price
Inquire For Price
Log in for availability
1 * 100 g
Product Details & Documents
Synonyms:  5-Nitro-2-pyridinol
  • CAS Number:5418-51-9
  • Formula:C₅H₄N₂O₃
  • MDL Number:MFCD00006276
  • Molecular Weight:140.1 g/mol
  • Boiling Pt:180…200 °C (0,2 mmHg)
  • Boiling Pt:180…200 °C (0.2 mmHg)
  • Melting Pt:191…192 °C
  • EINECS:226-525-7
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.