Order Entry
Germany
Orders LinkContactUsLinkComponent
2-Hydroxy-5-nitropyridine ≥98%

2-Hydroxy-5-nitropyridine ≥98%

2-Hydroxy-5-nitropyridine ≥98%
Certificates
2-Hydroxy-5-nitropyridine ≥98%
2-Hydroxy-5-nitropyridine ≥98%

2 Options Available Below

Product Details & Documents

Purity: 98+% CAS No.: 5418-51-9 Molecular Formula: C5H4N2O3 Formula Weight: 140.10 Melting Point: 188-191° Beilstein Reference: 120352

Synonyms:  5-Nitro-2-pyridinol
  • CAS Number:5418-51-9
  • Formula:C₅H₄N₂O₃
  • MDL Number:MFCD00006276
  • Molecular Weight:140.1 g/mol
  • Boiling Pt:180…200 °C (0.2 mmHg)
  • Melting Pt:188…191 °C
  • Storage Temperature:Ambient
  • EINECS:226-525-7
Product Family Options

Product Information

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.