2-Isobutyl-3-methoxypyrazine, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:24683-00-9
:
Formula:C₉H₁₄N₂O
MDL Number:MFCD00006128
:166.22 g/mol
Boiling Pt:237 °C (1013 hPa)
Melting Pt:107 °C
Density:0.99 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
EINECS:246-402-1