2-Isobutyl-3-methoxypyrazine, Sigma-Aldrich®
Catalog # SIAL297666-1G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:166.22 g/mol
Boiling Pt:237 °C (1013 hPa)
Melting Pt:107 °C
Density:0.99 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
EINECS:246-402-1
CAS Number:24683-00-9
Formula:C₉H₁₄N₂O
MDL Number:MFCD00006128