2-Isobutyl-3-methoxypyrazine
Catalog # EHERC14394700
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:0,1 g
- Seq:3020
Specifications
About this item
:
:166.22 g/mol
Boiling Pt:237 °C (1013 hPa)
Boiling Pt:237 °C (1013 hPa)
Melting Pt:107 °C
Density:0.99 g/cm³ (20 °C)
EINECS:246-402-1
CAS Number:24683-00-9
Formula:C₉H₁₄N₂O
MDL Number:MFCD00006128