2-Isobutyl-3-methoxypyrazine
Supplier: CHEMPUR
Certificates
About this item
CAS Number:24683-00-9
:
Formula:C₉H₁₄N₂O
MDL Number:MFCD00006128
:166.22 g/mol
Boiling Pt:237 °C (1013 hPa)
Melting Pt:107 °C
Density:0.99 g/cm³ (20 °C)
EINECS:246-402-1