Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Ethyl-3-methoxypyrazine
2-Ethyl-3-methoxypyrazine
2-Ethyl-3-methoxypyrazine
2-Ethyl-3-methoxypyrazine
Supplier:
FLUOROCHEM
Certificates
Expand image
2-Ethyl-3-methoxypyrazine
Supplier:
FLUOROCHEM
About this item
CAS Number:
25680-58-4
:
Formula:
C₇H₁₀N₂O
MDL Number:
MFCD00006129
:
138.17 g/mol
Boiling Pt:
81…85 °C (20 mmHg)
Density:
1,04 g/cm³ (20 °C)
EINECS:
247-184-0