2-Methoxypyrazine, Sigma-Aldrich®
SIAL291420-5G
: Merck
:
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
:
:110.12 g/mol
:147…149 °C (1013 hPa)
:1.143 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
:221-579-8
:3149-28-8
:C₅H₆N₂O
:MFCD00006126