
Methylene blue Reag. Ph. Eur., Supelco®
Supplier: Merck
Certificates
About this item
CAS Number:61-73-4
:MFCD00012111
Formula:C₁₆H₁₈ClN₃S
MDL Number:MFCD00150008
:319.86 g/mol
Merck Index:12,06137
Melting Pt:180 °C
Density:1.757 g/cm³ (22 °C)
:M-Clarity™ quality level = MQ200
EINECS:200-515-2
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Solubility:50 g/l (20 °C)
- Melting Point:180 °C (decomposition)
- Molar Mass:319.86 g/mol (anhydrous)
- Bulk Density:400 - 600 kg/m3
- pH:3 (10 g/l, H2O, 20 °C)
Specification Test Results
Related Information
Certificates
Search by Lot NumberMERC1.59270.0010_TDS_EN MERC1.59270.0100_TDS_EN ProductPage_159270
Product specification
msds_159270