Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Methylene blue
Methylene blue, Millipore®
Methylene blue, Millipore®
Methylene blue, Millipore®
457250-1
:
Merck Millipore (Calbiochem)
:
Methylene blue, Millipore®
457250-1
:
Merck Millipore (Calbiochem)
:
Pack type:
Glass bottle for solids
Pk:
1 g
Seq:
3047
:
:
319.86 g/mol
:
12,06137
:
180 °C
:
1.757 g/cm³ (22 °C)
:
200-515-2
:
GHS07
:
Ambient
:
61-73-4
:
MFCD00012111
:
C₁₆H₁₈ClN₃S
:
MFCD00150008