1,3-Propanesultone
Catalog # MOLEM80404953
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:122.14 g/mol
Boiling Pt:298 °C (1013 hPa)
Melting Pt:31 °C
Density:1.3996 g/cm³ (35 °C)
EINECS:214-317-9
UN:2811
ADR:6.1,III
:GHS08
:GHS07
CAS Number:1120-71-4
Formula:C₃H₆O₃S
MDL Number:MFCD00005355