Order Entry
Germany
ContactUsLinkComponent
Silver hexafluoroantimonate ≥98%
Silver hexafluoroantimonate ≥98%
Catalog # STRE93-4748-1G
CAS Number:  
Silver hexafluoroantimonate ≥98%
Catalog # STRE93-4748-1G
CAS Number:  

Specifications

  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :343.62
Density:4.65…4.85 g/cm³ (20 °C)
EINECS:247-429-1
UN:3260
ADR:8,II
 :GHS05
 :GHS09
CAS Number:26042-64-8
Formula:AgF₆Sb
MDL Number:MFCD00003401