Order Entry
Germany
ContactUsLinkComponent
6-Hydroxy-2-naphthoic acid 98%
Irritant
6-Hydroxy-2-naphthoic acid 98%
Certificates
undefined
Irritant
6-Hydroxy-2-naphthoic acid 98%

Specification Test Results

  • Appearance:
    Light brown Powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    240 to 250 °C
  • Titration with TBAH:
    ≥97.5 %

Specification Test Results

Related Information