0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Germany
ContactUsLinkComponent
3-Hydroxy-2-naphthoic acid
3-Hydroxy-2-naphthoic acid ≥98%
3-Hydroxy-2-naphthoic acid ≥98%
:
Thermo Fisher Scientific
3-Hydroxy-2-naphthoic acid ≥98%
:
Thermo Fisher Scientific
:
92-70-6
:
:
C₁₁H₈O₃
:
MFCD00004103
:
188.18 g/mol
:
14,4835
:
368 °C (1013 hPa)
:
218…222 °C
:
220 °C (428 °F)
:
Ambient
:
202-180-8
:
GHS06
:
GHS07
msds_ALFAA19214
Organic Building Blocks and Intermediates
Thermo Scientific Acroseal 25 Years