Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Hydroxy-1-naphthoic acid
2-Hydroxy-1-naphthoic acid, Sigma-Aldrich®
2-Hydroxy-1-naphthoic acid, Sigma-Aldrich®
2-Hydroxy-1-naphthoic acid, Sigma-Aldrich®
Supplier:
Merck
Certificates
Expand image
2-Hydroxy-1-naphthoic acid, Sigma-Aldrich®
Supplier:
Merck
About this item
CAS Number:
2283-08-1
:
Formula:
C₁₁H₈O₃
MDL Number:
MFCD00004009
:
188.18 g/mol
:
M-Clarity™ quality level = MQ100
EINECS:
218-919-2