Di-tert-butyl disulphide, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:110-06-5
:
Formula:C₈H₁₈S₂
MDL Number:MFCD00008838
:178.36 g/mol
Boiling Pt:198 °C (1013 hPa)
Melting Pt:–5 °C
Density:0.9229 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:203-734-1