Ammonium hexafluorophosphate, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:16941-11-0
:
Formula:H₄F₆NP
MDL Number:MFCD00064642
:163 g/mol
Merck Index:12,00557
Melting Pt:198 °C
Density:2.18 g/cm³ (18 °C)
EINECS:241-009-1
ADR:8,III
:M-Clarity™ quality level = MQ100
UN:3260