
Ammonium hexafluorophosphate ≥99.5%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:16941-11-0
:
Formula:H₄F₆NP
MDL Number:MFCD00064642
:163 g/mol
Merck Index:14,526
Melting Pt:198 °C
Density:2.180
Storage Temperature:Ambient
EINECS:241-009-1
UN:3260
ADR:8,III
:GHS05