


Cetrimonium chloride 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:112-02-7
:
Formula:C₁₉H₄₂ClN
MDL Number:MFCD00011773
:320 g/mol
Melting Pt:234 °C
Density:0,9 g/cm³ (20 °C)
EINECS:203-928-6
UN:2923
ADR:8,III
:GHS05
:GHS06
:GHS09
Storage Temperature:Ambient
Specification Test Results
- Appearance:White to cream Powder
- Infrared spectrum:Conforms
- Titration Argentometric:≥98.5 %
- GC:≥94 %
- Specific optical rotation:-33.0° to -37.0° (20°C, 589 nm) (c=9 in CHCl3)