

Dodecyltrimethylammonium chloride for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:112-00-5
:
Formula:C₁₅H₃₄ClN
MDL Number:MFCD00041974
:263.89 g/mol
Melting Pt:248…249.5 °C
Storage Temperature:Ambient
EINECS:203-927-0
UN:3077
ADR:9,III
:M-Clarity™ quality level = MQ200
:GHS07
:GHS09
Specification Test Results
- Melting Point:237 °C (decomposition)
- Molar Mass:263.9 g/mol
- Hazard Class:6.1
- Hazard Class Description:Toxic Materials