Order Entry
Germany
ContactUsLinkComponent
Bismuth(III) nitrate basic, EMSURE® Reag. Ph. Eur. for analysis, Supelco®
Bismuth(III) nitrate basic, EMSURE® Reag. Ph. Eur. for analysis, Supelco®
Catalog # 1.01878.0100
Supplier:  Merck
CAS Number:  
Bismuth(III) nitrate basic, EMSURE® Reag. Ph. Eur. for analysis, Supelco®
Catalog # 1.01878.0100
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    100 g
  • Seq:
    3401

Specifications

Specification Test Results

  • Solubility:
    (20 °C) practically insoluble
  • Molar Mass:
    1461.99 g/mol
  • Bulk Density:
    600 kg/m3
  • Density:
    4.93 g/cm3 (20 °C)
  • pH:
    3.2 (50 g/l, H2O, 20 °C) (slurry)
  • Hazard Class:
    5.1
  • Hazard Class Description:
    Oxidizer

Specification Test Results

About this item

 :
 :1461.99 g/mol
Density:4.928 g/cm³ (20 °C)
Storage Temperature:Ambient
UN:1477
ADR:5.1,II
 :M-Clarity™ quality level = MQ300
EINECS:215-136-8
 :GHS03
 :GHS07
CAS Number:1304-85-4
Formula:Bi₅O(OH)₉(NO₃)₄
MDL Number:MFCD00064844