Order Entry
Germany
ContactUsLinkComponent
Sand
Sand
Catalog # UPTI75035A
Supplier:  Uptima
CAS Number:  
Sand
Catalog # UPTI75035A
Supplier:  Uptima
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    50 mg
  • Seq:
    2370

Specifications

About this item

 :
 :60.08 g/mol
Melting Pt:∼1700 °C
Density:∼2.6 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-545-4
CAS Number:7631-86-9
Formula:SiO₂
MDL Number:MFCD02100519