Sand 40-100 mesh, pure
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:14808-60-7
:7631-86-9
Formula:SiO₂
MDL Number:MFCD00011232
:60.08 g/mol
Boiling Pt:2230 °C
Melting Pt:1610 °C
Density:~ 2,6 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-545-4
Specification Test Results
- Appearance:Light beige to grey Crystals
- Additional info:typical chemical analysis:
- Particle size:40 to 100 mesh