Order Entry
Germany
ContactUsLinkComponent
Boron carbide ≥99.5% (metals basis), sputtering target, Ø 76.2 mm (3.0 in), Thickness: 3.18 mm (0.125 in)
Boron carbide ≥99.5% (metals basis), sputtering target, Ø 76.2 mm (3.0 in), Thickness: 3.18 mm (0.125 in)
Catalog # 41050.KS
CAS Number:  
Boron carbide ≥99.5% (metals basis), sputtering target, Ø 76.2 mm (3.0 in), Thickness: 3.18 mm (0.125 in)
Catalog # 41050.KS
CAS Number:  

Specifications

  • Pk:
    1
  • Seq:
    0001

Specifications

About this item

CAS Number:12069-32-8
 :
Formula:CB₄
MDL Number:MFCD00011520
 :55.25 g/mol
Merck Index:12,01374
Boiling Pt:∼3500 °C (1013 hPa)
Melting Pt:2350 °C
Density:2.52 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:235-111-5