Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Strontium titanate
Strontium titanate, Sigma-Aldrich®
Strontium titanate, Sigma-Aldrich®
Strontium titanate, Sigma-Aldrich®
SIAL634689-1EA
:
Merck
:
Strontium titanate, Sigma-Aldrich®
SIAL634689-1EA
:
Merck
:
Pk:
1
Seq:
0001
:
:
183.5 g/mol
:
2080 °C
:
5.11 g/cm³ (20 °C)
:
M-Clarity™ quality level = MQ100
:
235-044-1
:
12060-59-2
:
O₃SrTi
:
MFCD00049554