Strontium titanate substrate 10x10x0.5 mm, polished one side 100 orientation
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:12060-59-2
:
Formula:O₃SrTi
MDL Number:MFCD00049554
:183.5 g/mol
Melting Pt:2080 °C
Density:5.11 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:235-044-1