Order Entry
Germany
Orders LinkContactUsLinkComponent
Magnesium oxide, rectangular tray, Length: 100mm, Width: 100mm, Height: 25.4mm

Magnesium oxide, rectangular tray, Length: 100mm, Width: 100mm, Height: 25.4mm

1
Catalog # 45791.KS
Magnesium oxide, rectangular tray, Length: 100mm, Width: 100mm, Height: 25.4mm
Catalog # 45791.KS
CAS Number:  1309-48-4
Magnesium oxide, rectangular tray, Length: 100mm, Width: 100mm, Height: 25.4mm
Magnesium oxide, rectangular tray, Length: 100mm, Width: 100mm, Height: 25.4mm

1

CAS Number:  1309-48-4
Price
1 010,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Synonyms:  Magnesia usta
  • CAS Number:1309-48-4
  • Formula:MgO
  • MDL Number:MFCD00011109
  • Molecular Weight:40.3 g/mol
  • Merck Index:13,05700
  • Boiling Pt:3600 °C (1013 hPa)
  • Melting Pt:2800 °C
  • Density:3.58 g/cm³ (25 °C)
  • EINECS:215-171-9
Specifications

Specifications

  • Pk:
    1

Specifications