Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Potassium titanate
Potassium titanate
Potassium titanate
:
Thermo Fisher Scientific
Potassium titanate
:
Thermo Fisher Scientific
:
12030-97-6
:
:
K₂O₃Ti
:
MFCD00058803
:
174.07 g/mol
:
1615 °C
:
3.1 g/cm³ (20 °C)
:
Ambient
:
234-748-6
msds_ALFAB20263
Thermo Scientific Acroseal 25 Years