Order Entry
Germany
ContactUsLinkComponent
 
 
 
Potassium pyroantimonate ≥99.0% for the precipitation of sodium, Fluka™
 
 
 
Potassium pyroantimonate ≥99.0% for the precipitation of sodium, Fluka™
 

 

  • Assay (as KSbO₃):
    Min. 99 %
  • Loss on ignition (600°C, 2 h):
    Max. 22 %
  • Application test:
    complying