Order Entry
Germany
ContactUsLinkComponent
 
Toxic
Corrosive
Lithium hexafluorophosphate
Catalog # FLUO002847-100G
Supplier:  FLUOROCHEM
CAS Number:  
Toxic
Corrosive
Lithium hexafluorophosphate
Catalog # FLUO002847-100G
Supplier:  FLUOROCHEM
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
Density:∼1.5 g/cm³ (20 °C)
EINECS:244-334-7
ADR:8,I
 :151.91 g/mol
UN:3260
 :GHS06
 :GHS05
Storage Temperature:Ambient
CAS Number:21324-40-3
Formula:F₆LiP
MDL Number:MFCD00011096