Industrial gloves, HyFlex® 11-528
Supplier: Ansell
Certificates
About this item
This glass-free INTERCEPT liner provides increased cut protection and lasting comfort and protection for hard-working hands.
- Skin friendly for increased comfort
- INTERCEPT Technology® for enhanced cut protection
- Enhanced flexibility for range of motion and tactility for handling small parts
- Grip performance for safe handling in dry to light oily applications
- NBR foam coating provides high grip and abrasion (ANSI 4)
- DMF-free
Ideal for use in the automotive and transportation industry.
Recommended for: Handling metal sheets & panels; handling parts with sharp, rough edges; final assembly; assembly & inspection of components; applying finish to materials.
Certifications: EN 388:2016 (3X32B); Cat. II
Food contact approved
Food contact approved
Packaging: 12 pairs per bag.
Specifications
- Catalog No:
- ANSE11-528/10
- ANSE11-528/8
- ANSE11-528/7
- ANSE11-528/6
- ANSE11-528/9
- ANSE11-528/11
- Ambidextrous / hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Liner material:
- HPPE/basalt/polyester/Spandex
- HPPE/basalt/polyester/Spandex
- HPPE/basalt/polyester/Spandex
- HPPE/basalt/polyester/Spandex
- HPPE/basalt/polyester/Spandex
- HPPE/basalt/polyester/Spandex
- Finish:
- Palm nitrile coated
- Palm nitrile coated
- Palm nitrile coated
- Palm nitrile coated
- Palm nitrile coated
- Palm nitrile coated
- Gauge:
- 18
- 18
- 18
- 18
- 18
- 18
- Cuff style:
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- EN norm:
- EN 388:2016 (3X32B)
- EN 388:2016 (3X32B)
- EN 388:2016 (3X32B)
- EN 388:2016 (3X32B)
- EN 388:2016 (3X32B)
- EN 388:2016 (3X32B)
- Anti-static:
- +
- +
- +
- +
- +
- +
- Food contact approved:
- +
- +
- +
- +
- +
- +