Industrial gloves, HyFlex® 11-738
Supplier: Ansell
Certificates
About this item
These tough gloves feature a unique combination of INTERCEPT Technology® yarn, Nylon, Lycra®, and glass fiber.
- Ultra strong fibers provide extreme resistance against cuts and burrs
- Reinforced thumb crotch for increased protection & extended use life
- INTERCEPT Cut Resistance Technology for extreme cut protection
- Made with water-based polyurethane for enhanced comfort and dexterity
- Silicone- and solvent-free
FORTIX™ Technology for extended product use life and increased protection with up to 12x more durability in the thumb crotch region versus unreinforced glove equivalents.
Recommended for: Handling of sharp-edged objects, pre-assembling, cutting of small dry or lightly oiled parts and maintenance; handling of glass sheets, sharpedged objects; handling of metal sheets, cutting of dry, painted or galvanised pieces.
Certifications: EN 388:2016 (4X42C); Cat. II
Oeko-Tex® 100 Certified
Oeko-Tex® 100 Certified
Packaging: 12 pairs per bag.
Specifications
- Catalog No:
- ANSE11-738/7
- ANSE11-738/10
- ANSE11-738/6
- ANSE11-738/8
- ANSE11-738/11
- ANSE11-738/9
- Ambidextrous / hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Latex / latex-free:
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Liner material:
- HPPE/glass fibre
- HPPE/glass fibre
- HPPE/glass fibre
- HPPE/glass fibre
- HPPE/glass fibre
- HPPE/glass fibre
- Finish:
- Palm PU/nitrile coated
- Palm PU/nitrile coated
- Palm PU/nitrile coated
- Palm PU/nitrile coated
- Palm PU/nitrile coated
- Palm PU/nitrile coated
- Gauge:
- 13
- 13
- 13
- 13
- 13
- 13
- Cuff style:
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- EN norm:
- EN 388:2016 (4X42C)
- EN 388:2016 (4X42C)
- EN 388:2016 (4X42C)
- EN 388:2016 (4X42C)
- EN 388:2016 (4X42C)
- EN 388:2016 (4X42C)
- Anti-static:
- +
- +
- +
- +
- +
- +