Industrial gloves, HyFlex® 11-300
Supplier: Ansell
Certificates
About this item
Glove liners made of high-elasticity polyamide for optimal fit.
- Glove knitted with a seamless non-stop process
- Excellent comfort and dexterity
- Ambidextrous to increase wear life
- Low-lint glove
Recommended for light duty tasks and food applications.
Certifications: Food contact approved
Packaging: 12 pairs per bag.
Specifications
- Catalog No:
- ANSE11-300/10
- ANSE11-300/11
- ANSE11-300/8
- ANSE11-300/7
- ANSE11-300/9
- Ambidextrous / hand-specific:
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Liner material:
- Polyamide
- Polyamide
- Polyamide
- Polyamide
- Polyamide
- Gauge:
- 13
- 13
- 13
- 13
- 13
- Cuff style:
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- Food contact approved:
- +
- +
- +
- +
- +