Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1-(4-Chlorophenyl)piperazine
1-(4-Chlorophenyl)piperazine, Sigma-Aldrich®
1-(4-Chlorophenyl)piperazine, Sigma-Aldrich®
1-(4-Chlorophenyl)piperazine, Sigma-Aldrich®
Supplier:
Merck
Certificates
Expand image
1-(4-Chlorophenyl)piperazine, Sigma-Aldrich®
Supplier:
Merck
About this item
CAS Number:
38212-33-8
:
Formula:
C₁₀H₁₃ClN₂
MDL Number:
MFCD00044823
:
196.68 g/mol
Boiling Pt:
137…138.5 °C (1 mmHg)
Melting Pt:
71.5…73.5 °C