1-(4-Chlorophenyl)piperazine
Catalog # FLUO009760-25G
Supplier: FLUOROCHEM
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:196.68 g/mol
Boiling Pt:137…138.5 °C (1 mmHg)
Melting Pt:71.5…73.5 °C
CAS Number:38212-33-8
Formula:C₁₀H₁₃ClN₂
MDL Number:MFCD00044823
Boiling Pt:137…138,5 °C (1 mmHg)
Melting Pt:71,5…73.5 °C