1-(3-Bromophenyl)piperazine
Marketsource
Supplier: FLUOROCHEM
Jump to:
About this item
- CAS Number:31197-30-5
- Formula:C₁₀H₁₃BrN₂
- MDL Number:MFCD03412123
- Molecular Weight:241.13 g/mol
2 Options Available Below
Product Details & Documents
- CAS Number:31197-30-5
- Formula:C₁₀H₁₃BrN₂
- MDL Number:MFCD03412123
- Molecular Weight:241.13 g/mol
Product Family Options
Product Information
- Pack typePkAvailabilityPrice
- Specifications:MarketsourceMarketsource
- Specifications:MarketsourceMarketsource
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



