Order Entry
Germany
Orders LinkContactUsLinkComponent
PIPES

PIPES

Marketsource
Supplier:   Molekula
PIPES
Supplier:  Molekula
Marketsource
Certificates
PIPES

2 Options Available Below

Product Details & Documents
Synonyms:  Piperazine-1,4-bis(2-ethanesulphonic acid), 2,2'-(Piperazine-1,4-diyl)diethanesulphonic acid, Piperazine-N,N'-bis-2-ethanesulphonic acid, 2,2'-(Piperazine-1,4-diyl)bis(ethanesulphonic) acid
  • CAS Number:5625-37-6
  • Formula:C₈H₁₈N₂O₆S₂
  • MDL Number:MFCD00006159
  • Molecular Weight:302.37 g/mol
  • Merck Index:12,07633
  • Boiling Pt:499 °C (1013 hPa)
  • Melting Pt:300 °C
  • Storage Temperature:Ambient
  • EINECS:227-057-6
Specifications for Product Family

Specifications

Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g

    Specifications:
    Marketsource
    More
    Specifications
    Cat. No.
    MOLE37406078-100G
    Marketsource
  • Glass bottle for solids
    500 g
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g

    Specifications:
    Marketsource
    More
    Specifications
    Cat. No.
    MOLE37406078-500G
    Marketsource
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.