Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
trans-1,4-Diaminocyclohexane
trans-1,4-Diaminocyclohexane, Sigma-Aldrich®
trans-1,4-Diaminocyclohexane, Sigma-Aldrich®
trans-1,4-Diaminocyclohexane, Sigma-Aldrich®
Supplier:
Merck
Certificates
Expand image
trans-1,4-Diaminocyclohexane, Sigma-Aldrich®
Supplier:
Merck
About this item
CAS Number:
2615-25-0
:
Formula:
C₆H₁₄N₂
MDL Number:
MFCD00075174
:
114.19 g/mol
Boiling Pt:
199 °C (1013 hPa)
Melting Pt:
70 °C