

Tetrachloro-p-benzoquinone
Catalog # MOLEM42579469
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:500 g
- Seq:3645
Specifications
About this item
:
:245.88 g/mol
Merck Index:13,02088
Boiling Pt:270 °C (1013 hPa)
Melting Pt:290…293 °C
Density:1.97 g/cm³ (20 °C)
Flash Pt:>100 °C
EINECS:204-274-4
:GHS07
:GHS09
Storage Temperature:Ambient
CAS Number:118-75-2
Formula:C₆Cl₄O₂
MDL Number:MFCD00001594